Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807130
Encompass
LumidoxII LumLamp, with 590-AMBER LEDs, 15 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-07130 Supplier Analytical Sales & Services Supplier No. LUM2L590C15
LumidoxII LumLamp, with 590-AMBER LEDs, 15 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens