Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807164
Encompass
LumidoxII LumLamp, with 630-RED LEDs, 40 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens
Catalog No. 50-008-07164 Supplier Analytical Sales & Services Supplier No. LUM2L630C40
LumidoxII LumLamp, with 630-RED LEDs, 40 Degree Extra Wide Beam Reflector and Clear Polycarbonate Lens