Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000807165
Encompass
LumidoxII LumLamp, with 590-AMBER LEDs, 30 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-07165 Supplier Analytical Sales & Services Supplier No. LUM2L590Q30
LumidoxII LumLamp, with 590-AMBER LEDs, 30 Degree Extra Wide Beam Reflector and Quartz Lens