Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805434
Encompass
LumidoxII LumLamp, with 445-INDIGO LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens
Catalog No. 50-008-05434 Supplier Analytical Sales & Services Supplier No. LUM2L445Q80
LumidoxII LumLamp, with 445-INDIGO LEDs, 80 Degree Extra Wide Beam Reflector and Quartz Lens