Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805438
Encompass
LumidoxII LumLamp, with UV365 LEDs, 15 Degree Narrow Spot Beam Reflector and Quartz Lens
Catalog No. 50-008-05438 Supplier Analytical Sales & Services Supplier No. LUM2L365Q15
Add to Cart
Add to Cart
LumidoxII LumLamp, with UV365 LEDs, 15 Degree Narrow Spot Beam Reflector and Quartz Lens