Learn More
Analytical Sales & Services Lumidox II LumLamp
Catalog No. 5000805440
Encompass
LumidoxII LumLamp, with 527-GREEN LEDs, 15 Degree Wide Beam Reflector and Diffused Polycarbonate Lens
Catalog No. 50-008-05440 Supplier Analytical Sales & Services Supplier No. LUM2L527D15
LumidoxII LumLamp, with 527-GREEN LEDs, 15 Degree Wide Beam Reflector and Diffused Polycarbonate Lens