Learn More
Analytical Sales & Services LumidoxII 48-Position LED Arrays for TCR - 527-Green
Supplier:  Analytical Sales & Services LUM248LS527
Encompass
LumidoxII 48 Well, 527-GREEN LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-07239