Learn More
Analytical Sales & Services LumidoxII 48-Position LED Arrays for TCR - 660-Deep Red
Catalog No. 5000805160
Encompass
LumidoxII 48 Well, 660-DEEP RED LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05160 Supplier Analytical Sales & Services Supplier No. LUM248LS660
LumidoxII 48 Well, 660-DEEP RED LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)