Learn More
Analytical Sales & Services LumidoxII 48-Position LED Arrays for TCR - 730-IR
Catalog No. 5000806379
Encompass
LumidoxII 48 Well, 730-IR LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06379 Supplier Analytical Sales & Services Supplier No. LUM248LS730
LumidoxII 48 Well, 730-IR LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)