Learn More
Analytical Sales & Services LumidoxII 48-Position LED Arrays for TCR - White
Catalog No. 5000805185
Encompass
LumidoxII 48 Well, WHITE LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05185 Supplier Analytical Sales & Services Supplier No. LUM248LSWHT
LumidoxII 48 Well, WHITE LED Array for TCR, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)