Learn More
LUMIGEN INCORPORATED LUMIGEN TMA -100
Catalog No. NC0240697
Encompass
Lumigen ECL Ultra (TMa-6) is a proprietary acridan based chemiluminescent substrate for ultra-sensitive detection of Western blotted and ELISa captured proteins that are bound to horseradish peroxidase (HRP) labeled antibodies.
Catalog No. NC0240697 Supplier LUMIGEN INCORPORATED Supplier No. TMA100
Lumigen ECL Ultra (TMa-6) is a proprietary acridan based chemiluminescent substrate for ultra-sensitive detection of Western blotted and ELISa captured proteins that are bound to horseradish peroxidase (HRP) labeled antibodies.