Learn More
Seracare Life Sciences Inc LUMIGLO HRP CHEMI SUBS
Supplier:  Seracare Life Sciences Inc 54300042
Encompass_Preferred
Non-isotopic, luminol-based chemiluminescent substrate designed for the rapid and sensitive detection of peroxidase labeled-conjugates. It provides increased sensitivity over chromogenic substrates in both blotting and ELISA applications. In blotting applications, permanent results are recorded on X-ray film. The use of LumiGLO allows for multiple stripping and reprobing of blots. In ELISA, positive reactions are read and rapidly detected using a luminometer. LumiGLO provides a dynamic range that is linear for a longer period of time than other chemiluminescent substrates. 2-Component format, simply mix and use. Safe to use: non-hazardous.
Catalog No. 50-675-10