Learn More
Seracare Life Sciences Inc LUMIGLO WESTERN BLOTTING KIT
Catalog No. 5067501
Encompass_Preferred
The Protein Detector LumiGLO Western Blot Kit is designed for low picogram detection of proteins captured on immunoblots. The combination of a highly specific, stable liquid conjugate and a sensitive, chemiluminescent substrate allows rapid and accurate identification of samples. All solutions required for blocking and washing the membrane and for diluting antibodies are provided. For added flexibility, affinity purified anti-mouse and anti-rabbit secondary antibodies conjugated with horseradish peroxidase are provided in stabilized, liquid form for quick dilution. LumiGLO Chemiluminescent Substrate is provided as a stable two component solution, which emits light when reacted with the HRP-labeled antibody.
This item is not returnable. View return policy
Catalog No. 50-675-01 Supplier Seracare Life Sciences Inc Supplier No. 54100009
The Protein Detector LumiGLO Western Blot Kit is designed for low picogram detection of proteins captured on immunoblots. The combination of a highly specific, stable liquid conjugate and a sensitive, chemiluminescent substrate allows rapid and accurate identification of samples. All solutions required for blocking and washing the membrane and for diluting antibodies are provided. For added flexibility, affinity purified anti-mouse and anti-rabbit secondary antibodies conjugated with horseradish peroxidase are provided in stabilized, liquid form for quick dilution. LumiGLO Chemiluminescent Substrate is provided as a stable two component solution, which emits light when reacted with the HRP-labeled antibody.