Learn More
Molecular Devices Molecular Devices Validation Plate, Absorbance, NIST-Traceable, ISO 17025 Recertifiable
Catalog No. 5000014360
Encompass_Preferred
The SpectraTest ABS2 Validation Plate from Molecular Devices is an automated absorbance validation tool used to verify the optical performance and sensitivity of absorbance-capable microplate readers. It provides NIST-traceable photometric checks and supports recertification to maintain GMP and GLP compliance; requires SoftMax Pro software version 6.5.1 or higher.
  • Validates absorbance sensitivity and photometric accuracy.
  • Automated certificate value entry for streamlined validation workflows.
  • Compatible with absorbance-capable microplate readers and automated handling systems.
  • Supports annual recertification with ISO 17025 traceability.
  • Designed for GMP and GLP instrument performance verification.
  • Factory recertification and repair services reduce downtime.
Catalog No. 50-000-14360 Supplier Molecular Devices Supplier No. 02006191
Add to Cart
Add to Cart
Request a Quote
The SpectraTest ABS2 Validation Plate from Molecular Devices is an automated absorbance validation tool used to verify the optical performance and sensitivity of absorbance-capable microplate readers. It provides NIST-traceable photometric checks and supports recertification to maintain GMP and GLP compliance; requires SoftMax Pro software version 6.5.1 or higher.
  • Validates absorbance sensitivity and photometric accuracy.
  • Automated certificate value entry for streamlined validation workflows.
  • Compatible with absorbance-capable microplate readers and automated handling systems.
  • Supports annual recertification with ISO 17025 traceability.
  • Designed for GMP and GLP instrument performance verification.
  • Factory recertification and repair services reduce downtime.