Learn More
Electron Microscopy Sciences Luminescent Thermometer, Model B
SDP
Catalog No. 5025289
Encompass_Preferred
Precision Dial Thermometer: easy to read Fahrenheit and Centigrade scales. Hermetically sealed; water proof; will not fog; made from corrosion resistant stainless steel. Comes with an adjustable positioning clamp. Accuracy to 1/2 degree. 2.5" luminescent dial with a 8" stem, 1/2 deg graduations and calibration adjustment.
Catalog No. 50-252-89 Supplier Electron Microscopy Sciences Supplier No. 74389
Precision Dial Thermometer: easy to read Fahrenheit and Centigrade scales. Hermetically sealed; water proof; will not fog; made from corrosion resistant stainless steel. Comes with an adjustable positioning clamp. Accuracy to 1/2 degree. 2.5" luminescent dial with a 8" stem, 1/2 deg graduations and calibration adjustment.