Learn More
Medchemexpress LLC Luminespib | 747412-49-3 | MFCD14635361 | 99.8% | 465.54 | C26H31N3O5 | 200 MG
SDP
Catalog No. 5000246347
Encompass_Preferred
Luminespib (VER-52296) is a third-generation heat shock protein 90 (HSP90) inhibitor supplied as a 200 mg powder for research and analytical applications. It shows low-nanomolar potency against HSP90α and HSP90β and is provided as a high-purity analytical standard.
Catalog No. 50-002-46347 Supplier Medchemexpress LLC Supplier No. HY10215200MG
Luminespib (VER-52296) is a third-generation heat shock protein 90 (HSP90) inhibitor supplied as a 200 mg powder for research and analytical applications. It shows low-nanomolar potency against HSP90α and HSP90β and is provided as a high-purity analytical standard.