Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luminespib | 747412-49-3 | MFCD14635361 | 99.8% | 465.54 g·mol⁻¹ | C26H31N3O5 | 1 ML
SDP

Catalog No. 5000246548
Encompass_Preferred

Luminespib is a small-molecule heat shock protein 90 (HSP90) inhibitor supplied as a 10 mM solution in DMSO for research applications. It demonstrates low-nanomolar potency against HSP90 isoforms and is used in biochemical and cellular studies investigating HSP90-related pathways.

  • Potent HSP90 inhibitor; IC50 7.8 nM (HSP90α) and 21 nM (HSP90β).
  • Supplied as a 10 mM solution in DMSO; 1 mL vial.
  • Molecular formula C26H31N3O5; molecular weight 465.54 g·mol⁻¹.
  • High purity suitable for research applications.
  • Typical storage at -20 °C to maintain stability.

Catalog No. 50-002-46548 Supplier Medchemexpress LLC Supplier No. HY1021510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luminespib is a small-molecule heat shock protein 90 (HSP90) inhibitor supplied as a 10 mM solution in DMSO for research applications. It demonstrates low-nanomolar potency against HSP90 isoforms and is used in biochemical and cellular studies investigating HSP90-related pathways.

  • Potent HSP90 inhibitor; IC50 7.8 nM (HSP90α) and 21 nM (HSP90β).
  • Supplied as a 10 mM solution in DMSO; 1 mL vial.
  • Molecular formula C26H31N3O5; molecular weight 465.54 g·mol⁻¹.
  • High purity suitable for research applications.
  • Typical storage at -20 °C to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.