Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ xMAP™ Sheath Fluid Concentrate (20X)
Description
- Delivery medium that carries sample to optics component of Luminex™ instruments
- Fluid is conveniently packaged as concentrated fluid (20X) that is diluted before use
Stability and Storage
- Store the unopened product at room temperature. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Concentration | 20X |
| Product Line | xMAP |
| Shipping Condition | Room Temperature |
For Laboratory Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?