Learn More
Sigma Aldrich Fine Chemicals Biosciences LuminoCtR qPCR ReadyMixTM
Catalog No. 502904816
Encompass
LuminoCt(R) qPCR ReadyMix combines the performance enhancements of our JumpStart(TM) Taq antibody for hot start PCR with the convenience of an easy-to-use reaction mixture. This is the ideal SOLUTIONon for performing high-throughput quantitative PCR methods that rely on a fluorescent probe.
Catalog No. 50-290-4816 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L6669100RXN
LuminoCt(R) qPCR ReadyMix combines the performance enhancements of our JumpStart(TM) Taq antibody for hot start PCR with the convenience of an easy-to-use reaction mixture. This is the ideal SOLUTIONon for performing high-throughput quantitative PCR methods that rely on a fluorescent probe.