Learn More
Medchemexpress LLC HY-15922 500mg Medchemexpress, Luminol CAS:521-31-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15922500MG
Encompass_Preferred
Medchemexpress, HY-15922 500mg Luminol CAS:521-31-3 Luminol is a chemical that exhibits chemiluminescence with pKa values of 6.74 and 15.1. Luminol exhibits chemiluminescence (CL) at 425 nm λmax. Luminol is commonly used in forensics as a diagnostic tool for the detection of blood stains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1833