Learn More
Cayman Chemical Luminol, 521-31-3, 5 g
SDP
Supplier:  Cayman Chemical 168035
Encompass_Preferred
A cyclic diacyl hydrazide that exhibits chemiluminescence upon oxidation; HRP combined with H2O2 causes luminescent peroxidation of luminol; can also be oxidized, and chemiluminesce, by compounds containing iron, copper, gold, and cyanide
Catalog No. NC1313833