Learn More
Enzo Life Sciences Luminol (5g). CAS: 521-31-3
SDP
Supplier:  Enzo Life Sciences ALX610002G005
Encompass_Preferred
Key reagent for generating luminescence. A reagent for the highly sensitive and selective detection of nitric oxide (NO) in biological samples by a luminol-H2O2 chemiluminescence method. Alternative name: 5-Amino-2,3-dihydro-1,4-phthalazinedione, 3-Aminophthalhydrazide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (50mg/ml) or in 10% ammonia (hot, 100mg/ml). Long Term Storage: Ambient.
Catalog No. 50-200-9593