Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Luminol
SCI_ED

Catalog No. S25395 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25395 5 g Solid
S25395A 1 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25395
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Produces chemiluminescence in presence of H2O2 for detection of nitrates.

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
InChI Key HWYHZTIRURJOHG-UHFFFAOYSA-N
Synonym luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
PubChem CID 10638
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)NNC2=O

Specifications

Color Yellow
Packaging Solid
Linear Formula C8H7N3O2
Quantity 5 g
Formula Weight 177.17
Chemical Name or Material Luminol
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.