Learn More
Thermo Scientific Chemicals Luminol and oxidizing solutions (each bottle)
Description
Used by biologists in cellular assays for the detection of copper, iron, and cyanides, as well as of specific proteins by western blot. For chemiluminecent HRP-based detection of western blotting systems. Microestimation of glucose and glucose oxidase using enzyme-induced chemiluminescence6,7 - Inhibitor of poly ADP Ribose Synthase8 - Demonstrations of the phenomenon of chemiluminescence, using different sensitizers to produce different colors of light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Luminol and oxidizing solutions (each bottle) |
| Quantity | 250 mL |
| Sensitivity | Light sensitive |
| Physical Form | Liquid |
| Solubility Information | It is soluble in most polar organic solvents but is insoluble in water. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.