Learn More
Santa Cruz Biotechnology LUMINOL REAGENT
Catalog No. NC9601786
Encompass
Western Blotting Luminol Reagent recommended for Western blotting enhanced chemiluminescence applications using Horseradish Peroxidase (HRP) conju-gated secondary antibodies Sufficient for 2 000cm2 membrane area
Catalog No. NC9601786 Supplier Santa Cruz Biotechnology Supplier No. SC2048
Western Blotting Luminol Reagent recommended for Western blotting enhanced chemiluminescence applications using Horseradish Peroxidase (HRP) conju-gated secondary antibodies Sufficient for 2 000cm2 membrane area