Learn More
Crescent Chemical Co Inc LUMINOL RESEARCH GRADE
SDP
Catalog No. 50247809
Encompass_Preferred
Luminol research grade For microdetermination of superoxide dismutase (2). Peroxidase reagent (1). Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 28085.02 CAS 521-31-3
Catalog No. 50-247-809 Supplier Crescent Chemical Co Inc Supplier No. SE28085.02
Luminol research grade For microdetermination of superoxide dismutase (2). Peroxidase reagent (1). Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 28085.02 CAS 521-31-3