Learn More
Enzo Life Sciences Luminol (ultra pure) (1g). CAS: 521-31-3
SDP
Catalog No. 502012635
Encompass_Preferred
Luminol is a luminescent peroxidase substrate that can be used as a chemiluminescent reagent for the determination of viable mammalian cells and bacteria. Luminol also has forensic application as a presumptive test for latent blood detection and is commonly employed in detection strategies by flow injection analysis, for pharmaceutical and environmental characterization. Chemiluminescent and electrochemiluminescent probes based upon luminal are beginning to merge with biochip and microarray development as a possible substitutes for the more wellestablished fluorescent probes. Wavelength Maxima: Excitation 355nm, Emission 411nm Alternative name: 3-Aminophthalhydrazide, (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2635 Supplier Enzo Life Sciences Supplier No. ENZ52354
Luminol is a luminescent peroxidase substrate that can be used as a chemiluminescent reagent for the determination of viable mammalian cells and bacteria. Luminol also has forensic application as a presumptive test for latent blood detection and is commonly employed in detection strategies by flow injection analysis, for pharmaceutical and environmental characterization. Chemiluminescent and electrochemiluminescent probes based upon luminal are beginning to merge with biochip and microarray development as a possible substitutes for the more wellestablished fluorescent probes. Wavelength Maxima: Excitation 355nm, Emission 411nm Alternative name: 3-Aminophthalhydrazide, (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.