Learn More
Electron Microscopy Sciences Luminox Low Foaming Neutral Cleaner 1 GL
SDP
Catalog No. 501898965
Encompass_Preferred
Low foaming, free rinsing neutral pH detergent can be used for manual or machine cleaning. It is a safe-handling detergent for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for aluminum and soft metal, medical devices, pharmaceutical equipments, alkaline sensitive labwares, and medical instruments, aluminum trays. Dilute 3:100 and it has a pH 7.0
Catalog No. 50-189-8965 Supplier Electron Microscopy Sciences Supplier No. 6051404
Low foaming, free rinsing neutral pH detergent can be used for manual or machine cleaning. It is a safe-handling detergent for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for aluminum and soft metal, medical devices, pharmaceutical equipments, alkaline sensitive labwares, and medical instruments, aluminum trays. Dilute 3:100 and it has a pH 7.0