Learn More
Cayman Chemical Lumiracoxib, Purity: ≥98%, CAS: 220991-20-8, 100 mg
SDP
Catalog No. 502939795
Encompass_Preferred
A selective inhibitor of COX-2 over COX-1 (IC50s = 0.13 and 67 µM, respectively); reduces increases in the levels of PGE2 induced by IL-1β in human dermal fibroblasts (IC50 = 0.14 µM), as well as in LPS-stimulated RAW 264.7 cells at 1-100 µM; decreases LPS-induced increases in the levels of PGE2 in a rat model of air pouch inflammation at 3 and 10 mg/kg; reduces M. tuberculosis-induced increases in hind paw volume and the radiological and histopathological severity of arthritic lesions in a rat model of chronic arthritis at 2 mg/kg
Catalog No. 50-293-9795 Supplier Cayman Chemical Supplier No. 27645100 MG
A selective inhibitor of COX-2 over COX-1 (IC50s = 0.13 and 67 µM, respectively); reduces increases in the levels of PGE2 induced by IL-1β in human dermal fibroblasts (IC50 = 0.14 µM), as well as in LPS-stimulated RAW 264.7 cells at 1-100 µM; decreases LPS-induced increases in the levels of PGE2 in a rat model of air pouch inflammation at 3 and 10 mg/kg; reduces M. tuberculosis-induced increases in hind paw volume and the radiological and histopathological severity of arthritic lesions in a rat model of chronic arthritis at 2 mg/kg