Learn More
Apexbio Technology LLC Lumiracoxib 220991-20-8 250mg
SDP
Catalog No. 5000089896
Encompass_Preferred
Lumiracoxib (CAS 220991-20-8) is a small molecule that selectively inhibits cyclooxygenase-2 (COX-2) an enzyme involved in prostaglandin synthesis and inflammatory signaling In biochemical assays lumiracoxib demonstrates an IC of 0 14 M and a K of 0 06 M for COX-2 with a selectivity ratio of 515-fold over cyclooxygenase-1 (COX-1) This high degree of isoform selectivity makes lumiracoxib a valuable tool for investigating the specific roles of COX-2 in inflammation and for exploring COX-2 targeted therapeutic strategies in inflammatory and pain-related disorders
Catalog No. 50-000-89896 Supplier Apexbio Technology LLC Supplier No. B1458250
Lumiracoxib (CAS 220991-20-8) is a small molecule that selectively inhibits cyclooxygenase-2 (COX-2) an enzyme involved in prostaglandin synthesis and inflammatory signaling In biochemical assays lumiracoxib demonstrates an IC of 0 14 M and a K of 0 06 M for COX-2 with a selectivity ratio of 515-fold over cyclooxygenase-1 (COX-1) This high degree of isoform selectivity makes lumiracoxib a valuable tool for investigating the specific roles of COX-2 in inflammation and for exploring COX-2 targeted therapeutic strategies in inflammatory and pain-related disorders