Learn More
Lj Star Inc LUMISTAR 1000
Catalog No. NC3793019
Encompass
LUMIUSL-33 LEDETL LISTED, UL 2108/CSA 22.2 CONFORMSNON-EXPLOSION PROOF LIGHT316 SS HOUSINGIP65: DUST AND WATERJET TIGHTPOWER: 100-240 VACWATTAGE: 12 WATT3 METER POWER CORD2 MIN/30 MIN INTERNAL TIMERPUSHBUTTON SWITCHFOR METACLAMP MOUNTSTAR 1000 > 100-240 VAC ETL VERSION
Catalog No. NC3793019 Supplier Lj Star Inc Supplier No. 33LSB6L4NNON
LUMIUSL-33 LEDETL LISTED, UL 2108/CSA 22.2 CONFORMSNON-EXPLOSION PROOF LIGHT316 SS HOUSINGIP65: DUST AND WATERJET TIGHTPOWER: 100-240 VACWATTAGE: 12 WATT3 METER POWER CORD2 MIN/30 MIN INTERNAL TIMERPUSHBUTTON SWITCHFOR METACLAMP MOUNTSTAR 1000 > 100-240 VAC ETL VERSION