Learn More
Medchemexpress LLC Lumisterol 3 | 5226-01-7 | 99.5% | 384.64 | 50 MG
SDP
Catalog No. 5000438580
Encompass_Preferred
Lumisterol 3 is a normal human secosteroid metabolite, a derivative of vitamin D3 photoisomers, primarily used in the preparation of vitamin D. It is intended for research use only.
  • Normal human secosteroid metabolite
  • Derivative of vitamin D3 photoisomer
  • Used in the preparation of vitamin D
  • Reduces UVB-induced cellular oxidant formation
  • Inhibits proliferation of UVB-irradiated keratinocytes
  • Promotes repair of cellular DNA damage induced by UVB irradiation
  • Increases nuclear/cytosolic Nrf2 ratio and nuclear p53 expression
  • Enhances expression of Nrf2-regulated antioxidant proteins such as catalase and HO-1
Catalog No. 50-004-38580 Supplier Medchemexpress LLC Supplier No. HY11202350MG
Lumisterol 3 is a normal human secosteroid metabolite, a derivative of vitamin D3 photoisomers, primarily used in the preparation of vitamin D. It is intended for research use only.
  • Normal human secosteroid metabolite
  • Derivative of vitamin D3 photoisomer
  • Used in the preparation of vitamin D
  • Reduces UVB-induced cellular oxidant formation
  • Inhibits proliferation of UVB-irradiated keratinocytes
  • Promotes repair of cellular DNA damage induced by UVB irradiation
  • Increases nuclear/cytosolic Nrf2 ratio and nuclear p53 expression
  • Enhances expression of Nrf2-regulated antioxidant proteins such as catalase and HO-1