Learn More
Greiner Bio-One LUMITRAC™ 200 Medium-Binding 384-well Microplates
Catalog No. 07001035 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-001-035 White
1 options
Catalog No. 07-001-035 Supplier Greiner Bio-One Supplier No. 78107525
Add to Cart
Add to Cart
Medium-binding microplates.
  • Made from polystyrene
  • Cell culture treated
  • With rounded-square wells
  • Flat solid bottom
Service Plan
No
Warranty and Services
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Binding Type Medium Binding
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 384
Well Shape Flat Bottom (F-well)
Quantity 150/Pk.