Learn More
Greiner Bio-One LUMITRAC™ 600 μClear™ Bottom High-Binding 96-well Microplate
Shop All Greiner Bio-One Products

Specifications
Specifications
| Barcode | Alphanumeric |
| Format | Standard |
| Lid | Without Lid |
| Detection Method | Luminescence |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Packaging | Inner bags of 10 |
| Binding Type | High Binding |
| Color | White, Clear Bottom |
| Product Type | Assay Microplate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.