Learn More
Greiner Bio-One LUMITRAC™ 600 High-Binding 384-well Microplates
Catalog No. 07001034 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-001-034 White
07-001-036 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-001-034 Supplier Greiner Bio-One Supplier No. 78107425
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High-binding microplates.
  • Made from polystyrene
  • Cell culture treated
  • With rounded-square wells
  • Flat solid bottom
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 16 x 24
Binding Type High Binding
Color White
Material Polystyrene
No. of Wells 384
Well Shape Flat Bottom (F-well)
Quantity 25/Pk.