Learn More
Greiner Bio-One LUMITRAC™ and FLUOTRAC™ Binding 96-well Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07000129 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
White
Surface Treatment:
FLUOTRAC 200
FLUOTRAC 600
LUMITRAC 200
LUMITRAC 600
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Surface Treatment Sterility
07-000-129 Black FLUOTRAC 200 Non-sterile
07-000-128 White LUMITRAC 600 Sterile
07-000-130 White LUMITRAC 200 Non-sterile
07-000-127 Black FLUOTRAC 600 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-129 Supplier Greiner Bio-One Supplier No. 655076
Add to Cart
Add to Cart
For luminescence measurements. Greiner Bio-One LUMITRAC™ and FLUOTRAC™ Binding 96-Well Polystyrene Microplates are specifically treated to provide an increased protein binding.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Sample carryover and cross-contamination risk is minimized
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color Black
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment FLUOTRAC 200
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney