Learn More
Cayman Chemical Lumogallion, Purity: ≥98%, CAS: 4386-25-8, 100 mg
SDP
Catalog No. 502909948
Encompass_Preferred
A fluorescent probe for aluminum; binds to aluminum, forming a complex that displays excitation/emission maxima of 500/590 nm, respectively; has been used to detect aluminum in natural waters and human urine and serum samples by HPLC, as well as in fixed soybean roots and THP-1 cells by confocal microscopy
Catalog No. 50-290-9948 Supplier Cayman Chemical Supplier No. 35596100 MG
A fluorescent probe for aluminum; binds to aluminum, forming a complex that displays excitation/emission maxima of 500/590 nm, respectively; has been used to detect aluminum in natural waters and human urine and serum samples by HPLC, as well as in fixed soybean roots and THP-1 cells by confocal microscopy