Learn More
Phenomenex Inc Luna 10 m C18(2) 100
Catalog No. 502529280 Shop All Phenomenex Products
Encompass
LC Column 250 x 4.6 mm
Catalog No. 50-252-9280 Supplier Phenomenex Inc Supplier No. 00G4253E0
Add to Cart
Add to Cart
LC Column 250 x 4.6 mm