Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 150 x 30 mm, AXIA™ Packed

Supplier:  Phenomenex Inc 00F4285U0AX

Encompass

Luna 10 m Phenyl-Hexyl 100A 00F-4285-U0-AX

Catalog No. 50-239-4129


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.