Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 250 x 30 mm, AXIA™ Packed

Supplier:  Phenomenex Inc 00G4324U0AX

Encompass

Luna 10 m PREP C18(2) 100 00G-4324-U0-AX

Catalog No. 50-239-6472


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.