Learn More
New England Biolabs, Inc. Luna Probe One-Step RT-qPCR 4X Mix with UDG - 1000 rxns
SDP
Catalog No. 502023996
Encompass
The Luna Probe One-Step RT-qPCR 4X Mix with UDG is designed for real-time detection of target RNA sequences using hydrolysis probes. In a single tube, RNA is first converted to cDNA by a reverse transcriptase, then a DNA-dependent DNA polymerase amplifies the cDNA, enabling quantitation via real time or quantitative PCR (qPCR). Probe-based qPCR/RT-qPCR monitors an increase in fluorescence upon 5' to 3' exonuclease cleavage of a quenched, target-specific probe to measure DNA amplification at each cycle of a PCR. At a point where the fluorescence signal is confidently detected over the background fluorescence, a quantification cycle or Cq value can be determined. Cq values can be used to evaluate relative target abundance between two or more samples.
Catalog No. 50-202-3996 Supplier New England Biolabs, Inc. Supplier No. M3019X
The Luna Probe One-Step RT-qPCR 4X Mix with UDG is designed for real-time detection of target RNA sequences using hydrolysis probes. In a single tube, RNA is first converted to cDNA by a reverse transcriptase, then a DNA-dependent DNA polymerase amplifies the cDNA, enabling quantitation via real time or quantitative PCR (qPCR). Probe-based qPCR/RT-qPCR monitors an increase in fluorescence upon 5' to 3' exonuclease cleavage of a quenched, target-specific probe to measure DNA amplification at each cycle of a PCR. At a point where the fluorescence signal is confidently detected over the background fluorescence, a quantification cycle or Cq value can be determined. Cq values can be used to evaluate relative target abundance between two or more samples.