Learn More
NEW ENGLAND BIOGROUP LUNA 2nd General Reusable Slide, Counts 1 sample at a time, Contains LUNA reusable slide and one glass coverslip, For brightfield and fluorescent cell counts.
SDP
Catalog No. NC3903966
Encompass
LUNA 2nd General Reusable Slide. Counts 1 sample at a time. Contains LUNA reusable slide and one glass coverslip. Can be used for brightfield and fluorescent cell counts. One year warranty
Catalog No. NC3903966 Supplier NEW ENGLAND BIOGROUP Supplier No. L12011
LUNA 2nd General Reusable Slide. Counts 1 sample at a time. Contains LUNA reusable slide and one glass coverslip. Can be used for brightfield and fluorescent cell counts. One year warranty