Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Luna 3 µm C8(2) 100 Å

Catalog No. 502394454 Shop All Phenomenex Products
Encompass

Luna 3 µm C8(2) 100 Å LC Column 150 x 3 mm

Catalog No. 50-239-4454 Supplier Phenomenex Inc Supplier No. 00F4248Y0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luna 3 µm C8(2) 100 Å LC Column 150 x 3 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.