Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 0.3 mm

Supplier:  Phenomenex Inc 00D4256AC

Encompass

Luna 3m Phenyl-Hexyl 100 00D-4256-AC

Catalog No. 50-239-5719


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.