Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Luna 3 µm Polar Pesticides

Catalog No. 502393888 Shop All Phenomenex Products
Encompass

Luna 3 µm Polar Pesticides LC Column 150 x 2.1 mm

Catalog No. 50-239-3888 Supplier Phenomenex Inc Supplier No. 00F4798AN
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luna 3 µm Polar Pesticides LC Column 150 x 2.1 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.