Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Luna 5 µm C18 100 Å

Catalog No. 502396100 Shop All Phenomenex Products
Encompass

Luna 5 µm C18 100 Å LC Column 50 x 2 mm

Catalog No. 50-239-6100 Supplier Phenomenex Inc Supplier No. 00B4041B0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luna 5 µm C18 100 Å LC Column 50 x 2 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.