Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 10 x 2 mm, MercuryMS™

Catalog No. 502395873 Shop All Phenomenex Products
Encompass

Luna 5 m Phenyl-Hexyl 100 00N-4257-B0-CE

Catalog No. 50-239-5873 Supplier Phenomenex Inc Supplier No. 00N4257B0CE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luna 5 m Phenyl-Hexyl 100 00N-4257-B0-CE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.