Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 2 mm

Catalog No. 502393151 Shop All Phenomenex Products
Encompass

Luna 5 m Silica (2) 100 00D-4274-B0

Catalog No. 50-239-3151 Supplier Phenomenex Inc Supplier No. 00D4274B0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luna 5 m Silica (2) 100 00D-4274-B0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.