Learn More
Phenomenex Inc Phenomenex HPLC Column, C18(2), 5 μm, 250 x 4.6 mm
Supplier:  Phenomenex Inc 00G4252EO
Encompass
Reversed-phase HPLC column designed for analytical separations that require high resolution and methylene selectivity. The 5 μm particle size and 100 Å pore size offer a balance of efficiency and loading capacity for small-molecule analyses, making the column suitable for USP methods and general LC method development.
  • High resolution and improved peak shape.
  • Methylene selectivity for challenging separations.
  • Consistent batch-to-batch performance.
  • Suitable for a wide range of reversed-phase methods.
  • Analytical scale 250 x 4.6 mm format for standard workflows.
Catalog No. NC3713214