Learn More
LOGOS BIOSYSTEMS INC 8-Channel Slides, 50/pk
SDP
Catalog No. NC1842174
Encompass
The disposable LUNA 8-Channel Slides provides much higer throughput of samples. Count 8 different samples at one go to increase your lab’s effieciency. With no mess or cleanup, these precision slides are convenient and maintain the highest standard of cell counting accuracy. The LUNA 8-Channel Slides are for brightfield and fluorescence cell counting and only compatible with the LUNA-FX7.
Catalog No. NC1842174 Supplier LOGOS BIOSYSTEMS INC Supplier No. L72001
The disposable LUNA 8-Channel Slides provides much higer throughput of samples. Count 8 different samples at one go to increase your lab’s effieciency. With no mess or cleanup, these precision slides are convenient and maintain the highest standard of cell counting accuracy. The LUNA 8-Channel Slides are for brightfield and fluorescence cell counting and only compatible with the LUNA-FX7.